Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139281.2 | NP_644810.1 | 3 | 519 | ||
GenBank mRNA | AF385437.1 | - | 3 | 519 | ||
AK095312.1 | - | 3 | 377 | |||
AL832494.1 | - | 2 | 325 | |||
BC133025.1 | - | 3 | 479 | 40147047 | ||
BC136517.1 | - | 3 | 479 | 9020507 |