Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAKMIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099433.1 | NP_001092903.1 | 4 | 1134 | ||
NM_001306133.1 | NP_001293062.1 | 4 | 924 | |||
NM_001306134.1 | NP_001293063.1 | 3 | 429 | |||
NM_144720.3 | NP_653321.1 | 4 | 1134 | |||
XM_011513400.2 | XP_011511702.1 | 3 | 603 | |||
XM_017007788.1 | XP_016863277.1 | 4 | 1098 | |||
XM_017007789.1 | XP_016863278.1 | 4 | 922 | |||
XM_017007790.1 | XP_016863279.1 | 4 | 1074 | |||
XM_017007791.1 | XP_016863280.1 | 4 | 1098 | |||
XM_017007792.1 | XP_016863281.1 | 4 | 1098 | |||
XM_017007793.1 | XP_016863282.1 | 4 | 1098 | |||
XM_017007794.1 | XP_016863283.1 | 4 | 1098 | |||
XM_017007795.1 | XP_016863284.1 | 4 | 1098 | |||
XM_017007796.1 | XP_016863285.1 | 4 | 1098 | |||
XM_017007797.1 | XP_016863286.1 | 3 | 603 | |||
XM_017007798.1 | XP_016863287.1 | 3 | 1241 | |||
XM_017007799.1 | XP_016863288.1 | 3 | 603 | |||
GenBank mRNA | AK056126.1 | - | 4 | 1098 | ||
AK098536.1 | - | 2 | 208 | |||
AK124718.1 | - | 4 | 922 | |||
AK294356.1 | - | 5 | 751 | |||
AK295339.1 | - | 3 | 429 | |||
AK307983.1 | - | 3 | 349 | |||
AM412307.1 | - | 4 | 1098 | |||
AM412308.1 | - | 4 | 1098 | |||
AM412309.1 | - | 4 | 1098 | |||
AM412310.1 | - | 3 | 603 | |||
AY382340.1 | - | 3 | 648 | |||
BC047075.1 | - | 4 | 854 | 5314069 |