Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153252.4 | NP_694984.4 | 15 | 1767 | ||
XM_005262113.3 | XP_005262170.1 | 15 | 1769 | |||
XM_017029384.1 | XP_016884873.1 | 11 | 1297 | |||
XM_017029385.1 | XP_016884874.1 | 15 | 1769 | |||
GenBank mRNA | AY497046.1 | - | 15 | 1767 | ||
AY497047.1 | - | 12 | 1699 | |||
AY497048.1 | - | 15 | 1790 | |||
AY497049.1 | - | 14 | 1760 | |||
AY497050.1 | - | 15 | 1740 | |||
AY497051.1 | - | 12 | 1327 | |||
AY497052.1 | - | 17 | 1926 | |||
AY497053.1 | - | 13 | 1515 | |||
AY497054.1 | - | 12 | 1416 | |||
AY497055.1 | - | 15 | 1701 | |||
AY497056.1 | - | 12 | 1672 | |||
AY497057.1 | - | 11 | 1571 | |||
AY497058.1 | - | 11 | 1410 | |||
AY497059.1 | - | 13 | 1781 | |||
AY497060.1 | - | 9 | 1348 | |||
BC111490.1 | - | 15 | 1717 |