Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172069.3 | NP_742066.2 | 2, 3 | 199 | ||
XM_017003352.1 | XP_016858841.1 | 2, 3 | 199 | |||
XM_017003353.1 | XP_016858842.1 | 2, 3 | 199 | |||
GenBank mRNA | AK126308.1 | - | 2, 3 | 199 | ||
AL832207.1 | - | 2, 3 | 192 | |||
BC063310.1 | - | 2, 3 | 226 | 30342490 |