Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004645.2 | NP_004636.1 | 2 | 304 | ||
GenBank mRNA | AK313616.1 | - | 2 | 286 | ||
BC010385.1 | - | 2 | 301 | 4275993 | ||
U06632.1 | - | 2 | 295 |