Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181715.2 | NP_859066.1 | 12 | 1747 | ||
XM_005244946.1 | XP_005245003.1 | 12 | 1743 | |||
XM_005244949.2 | XP_005245006.1 | 12 | 1620 | |||
XM_017000575.1 | XP_016856064.1 | 11 | 1516 | |||
XM_017000576.1 | XP_016856065.1 | 11 | 1501 | |||
XM_017000577.1 | XP_016856066.1 | 11 | 1645 | |||
XM_017000578.1 | XP_016856067.1 | 11 | 1630 | |||
XM_017000579.1 | XP_016856068.1 | 6 | 1042 | |||
GenBank mRNA | AF289574.1 | - | 3 | 706 | ||
AK096382.1 | - | 10 | 1338 | |||
AK223559.1 | - | 12 | 1700 | |||
AY360172.1 | - | 12 | 1718 | |||
AY726597.1 | - | 12 | 1527 | |||
BC033304.1 | - | 10 | 1349 | |||
BC053562.1 | - | 12 | 1658 | 6188068 |