Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190458.1 | NP_001177387.1 | 5 | 529 | ||
NM_001193536.1 | NP_001180465.1 | 5 | 529 | |||
NM_203447.3 | NP_982272.2 | 6 | 744 | |||
XM_011518045.2 | XP_011516347.1 | 6 | 599 | |||
XM_011518046.2 | XP_011516348.1 | 6 | 642 | |||
XM_011518047.2 | XP_011516349.1 | 6 | 825 | |||
XM_011518048.2 | XP_011516350.1 | 5 | 564 | |||
XM_017015173.1 | XP_016870662.1 | 6 | 599 | |||
XM_017015174.1 | XP_016870663.1 | 6 | 679 | |||
GenBank mRNA | AB191037.1 | - | 6 | 744 | ||
AK093572.1 | - | 5 | 527 | |||
AK310555.1 | - | 5 | 511 | |||
AL832270.1 | - | 5 | 493 | |||
BC045629.1 | - | 5 | 518 | |||
BC130518.1 | - | 5 | 507 | 40146549 | ||
BC143929.1 | - | 5 | 507 | 9052447 | ||
BU532484.1 | - | 5 | 532 | |||
DB147029.1 | - | 4 | 405 | |||
HM991480.1 | - | 5 | 525 | |||
KC736820.1 | - | 4 | 428 |