Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMSAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297707.1 | NP_001284636.1 | 2 | 593 | ||
NM_001297708.1 | NP_001284637.1 | 2 | 593 | |||
NM_203459.2 | NP_982284.1 | 2 | 593 | |||
XM_005245041.3 | XP_005245098.1 | 2 | 450 | |||
GenBank mRNA | AK095640.1 | - | 2 | 593 | ||
BC125229.1 | - | 2 | 372 | 40124603 | ||
BC125230.1 | - | 2 | 372 | 40124604 | ||
CA388618.1 | - | 2 | 273 |