Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMSAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297707.1 | NP_001284636.1 | 12 | 2255 | ||
NM_001297708.1 | NP_001284637.1 | 11 | 2174 | |||
NM_203459.2 | NP_982284.1 | 11 | 2222 | |||
XM_005245041.3 | XP_005245098.1 | 12 | 2064 | |||
XM_017000799.1 | XP_016856288.1 | 11 | 2648 | |||
GenBank mRNA | AB029001.2 | - | 11 | 1613 | ||
AK095640.1 | - | 11 | 2222 | |||
AK126821.1 | - | 11 | 1627 | |||
BC029449.1 | - | 1 | 214 | |||
BC056910.1 | - | 1 | 766 | |||
BC065508.1 | - | 1 | 573 | |||
BC125229.1 | - | 12 | 2034 | 40124603 | ||
BC125230.1 | - | 11 | 1953 | 40124604 |