Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206996.2 | NP_996879.1 | 6 | 813 | ||
XM_006710426.3 | XP_006710489.1 | 6 | 813 | |||
XM_011540934.1 | XP_011539236.1 | 6 | 813 | |||
XM_011540935.2 | XP_011539237.1 | 5 | 652 | |||
GenBank mRNA | AK091816.1 | - | 6 | 799 | ||
AY555274.1 | - | 6 | 799 | |||
BX648110.1 | - | 6 | 814 |