Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278211.1 | NP_001265140.1 | 5 | 2117 | ||
NM_001278212.1 | NP_001265141.1 | 5 | 2062 | |||
NM_001278213.1 | NP_001265142.1 | 4 | 1861 | |||
NM_001278214.1 | NP_001265143.1 | 4 | 1917 | |||
NM_018205.3 | NP_060675.1 | 4 | 1894 | |||
NM_018239.3 | NP_060709.2 | 4 | 1912 | |||
NM_207119.2 | NP_997002.1 | 5 | 2375 | |||
NR_103467.1 | 3 | 1767 | ||||
NR_103468.1 | 3 | 1749 | ||||
XM_017016374.1 | XP_016871863.1 | 5 | 2057 | |||
XM_017016375.1 | XP_016871864.1 | 5 | 2052 | |||
GenBank mRNA | AK001613.1 | - | 4 | 1871 | ||
AK001706.1 | - | 4 | 1886 | |||
AK094734.1 | - | 5 | 2058 | |||
AL122117.1 | - | 2 | 1599 | |||
AY358318.1 | - | 5 | 2116 | |||
BC024001.2 | - | 5 | 2053 | 4822655 |