Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256478.1 | NP_001243407.1 | 4 | 1130 | ||
GenBank mRNA | AK127983.1 | - | 6 | 1317 | ||
BC132821.1 | - | 4 | 1130 | 40146843 | ||
BC144202.1 | - | 4 | 1130 | 9052722 |