Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004699.3 | NP_004690.1 | 1, 2 | 229 | ||
GenBank mRNA | AD001530.1 | - | 1, 2 | 194 | ||
AK293119.1 | - | 1, 2 | 183 | |||
AK313697.1 | - | 1, 2 | 177 | |||
AL365368.1 | - | 1, 2 | 109 | |||
AU143518.1 | - | 1, 2 | 217 | |||
BC000028.2 | - | 1, 2 | 200 | 3505365 | ||
CR992165.1 | - | 1, 2 | 229 | |||
D83260.1 | - | 1, 2 | 99 |