Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513603.2 | XP_011511905.1 | 9 | 3776 | ||
XM_011513604.2 | XP_011511906.1 | 10 | 4042 | |||
XM_017008891.1 | XP_016864380.1 | 11 | 4575 | |||
XM_017008892.1 | XP_016864381.1 | 11 | 4575 | |||
XM_017008893.1 | XP_016864382.1 | 10 | 4408 | |||
GenBank mRNA | AK128339.1 | - | 7 | 1180 | ||
BC140710.1 | - | 7 | 1156 | 9021578 |