Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFRAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207410.2 | NP_997293.2 | 4, 5 | 452 | ||
GenBank mRNA | AY358198.1 | - | 4, 5 | 453 |