Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGGTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004582.3 | NP_004573.2 | 6 | 642 | ||
NR_073562.1 | 8 | 837 | ||||
GenBank mRNA | AB209023.1 | - | 5 | 564 | ||
AK128861.1 | - | 4 | 3245 | |||
AK293057.1 | - | 8 | 837 | |||
BC020790.1 | - | 6 | 635 | 4290610 | ||
CR456941.1 | - | 5 | 564 | |||
U49245.1 | - | 5 | 564 | |||
X98001.1 | - | 5 | 588 | |||
Y08201.1 | - | 5 | 567 |