Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARRES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004585.4 | NP_004576.2 | 3 | 367 | ||
GenBank mRNA | AB030815.1 | - | 3 | 323 | ||
AB453252.1 | - | 2 | 299 | |||
AF060228.1 | - | 3 | 328 | |||
AF092922.1 | - | 3 | 360 | |||
AK293357.1 | - | 3 | 352 | |||
AK312110.1 | - | 3 | 352 | |||
BC009678.1 | - | 3 | 347 | 3877277 | ||
DA973023.1 | - | 3 | 369 | |||
R55646.1 | - | 3 | 339 |