Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207363.2 | NP_997246.2 | 6 | 666 | ||
NM_207481.3 | NP_997364.3 | 6 | 666 | |||
XM_005263659.3 | XP_005263716.1 | 5 | 1248 | |||
XM_005263660.3 | XP_005263717.1 | 5 | 763 | |||
XM_011511097.2 | XP_011509399.1 | 7 | 1436 | |||
XM_011511098.2 | XP_011509400.1 | 7 | 1433 | |||
XM_011511099.2 | XP_011509401.1 | 6 | 831 | |||
XM_011511100.2 | XP_011509402.1 | 6 | 929 | |||
XM_011511101.1 | XP_011509403.1 | 4 | 339 | |||
XM_011511102.2 | XP_011509404.1 | 5 | 4765 | |||
XM_011511103.2 | XP_011509405.1 | 3 | 567 | |||
XM_011511104.1 | XP_011509406.1 | 3 | 218 | |||
XM_011511105.2 | XP_011509407.1 | 3 | 1009 | |||
XM_017003974.1 | XP_016859463.1 | 6 | 559 | |||
XM_017003975.1 | XP_016859464.1 | 5 | 861 | |||
XM_017003976.1 | XP_016859465.1 | 7 | 1457 | |||
XM_017003977.1 | XP_016859466.1 | 4 | 1367 | |||
XM_017003979.1 | XP_016859468.1 | 6 | 1306 | |||
XM_017003980.1 | XP_016859469.1 | 3 | 567 | |||
GenBank mRNA | AK057980.1 | - | 6 | 666 | ||
AK092189.1 | - | 4 | 341 | |||
AY946007.1 | - | 1 | 88 | |||
AY946008.1 | - | 1 | 88 | |||
BC110831.1 | - | 4 | 279 |