Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297738.1 | NP_001284667.1 | 2, 3 | 271 | ||
NM_004607.2 | NP_004598.1 | 2, 3 | 271 | |||
GenBank mRNA | AF038952.1 | - | 2, 3 | 216 | ||
AK300031.1 | - | 2, 3 | 193 | |||
AK312207.1 | - | 2, 3 | 239 | |||
BC018210.1 | - | 2, 3 | 256 | 3861138 | ||
BC105285.1 | - | 2, 3 | 207 | 6702573 | ||
BT007354.1 | - | 2, 3 | 167 | |||
CR542170.1 | - | 2, 3 | 167 | |||
CR542186.1 | - | 2, 3 | 167 | |||
DB538817.1 | - | 34 | ||||
DC312139.1 | - | 2, 3 | 271 |