Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123499.1 | - | 4 | 659 | ||
BC118595.1 | - | 2 | 352 | |||
BC122559.1 | - | 2 | 352 | |||
BC128606.1 | - | 3 | 563 | 40108149 |