Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plscr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195084.1 | NP_001182013.1 | 8 | 1242 | ||
NM_008880.3 | NP_032906.2 | 7 | 1237 | |||
GenBank mRNA | AF015790.1 | - | 8 | 1008 | ||
AK002410.1 | - | 7 | 1124 | |||
BC002012.1 | - | 8 | 1108 | 3490475 | ||
BC011299.1 | - | 7 | 1119 | 3595010 |