Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133393.1 | - | 14 | 1830 | ||
BC046617.1 | - | 15 | 1662 | |||
M81475.1 | - | 14 | 1803 |