Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008948.2 | NP_032974.2 | 6 | 798 | ||
GenBank mRNA | AK002542.1 | - | 6 | 518 | ||
AK083043.1 | - | 760 | ||||
AK150189.1 | - | 6 | 734 | |||
AK151370.1 | - | 6 | 518 | |||
AK152018.1 | - | 6 | 537 | |||
AK167002.1 | - | 6 | 746 | |||
AK167061.1 | - | 6 | 743 | |||
AK168263.1 | - | 5 | 467 | |||
AK168616.1 | - | 6 | 518 | |||
AK214668.1 | - | 1 | 20 | |||
BC005783.1 | - | 6 | 687 | 3603053 | ||
D49686.1 | - | 6 | 742 |