Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008959.3 | NP_032985.2 | 1 | 784 | ||
GenBank mRNA | AF042731.1 | - | 1 | 199 | ||
AK036990.1 | - | 1 | 246 | |||
AK054101.1 | - | 1 | 223 | |||
AK085857.1 | - | 1 | 785 | |||
AK086136.1 | - | 1 | 399 | |||
AK088048.1 | - | 1 | 266 | |||
AK137681.1 | - | 1 | 454 | |||
AK172873.1 | - | 1 | 761 | |||
BC003260.1 | - | 1 | 421 | 3501141 |