Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008972.2 | NP_032998.1 | 5 | 534 | ||
GenBank mRNA | AK037980.1 | - | 5 | 545 | ||
AK084218.1 | - | 5 | 544 | |||
AK088005.1 | - | 5 | 545 | |||
AK132003.1 | - | 5 | 551 | |||
AK142076.1 | - | 5 | 550 | |||
AK143576.1 | - | 5 | 545 | |||
AK145650.1 | - | 5 | 545 | |||
AK146454.1 | - | 5 | 544 | |||
AK146651.1 | - | 5 | 545 | |||
AK146849.1 | - | 5 | 545 | |||
AK151289.1 | - | 5 | 544 | |||
AK151511.1 | - | 5 | 548 | |||
AK151768.1 | - | 5 | 545 | |||
AK152032.1 | - | 5 | 544 | |||
AK153178.1 | - | 5 | 545 | |||
AK153496.1 | - | 5 | 545 | |||
AK153953.1 | - | 5 | 549 | |||
AK159128.1 | - | 5 | 545 | |||
AK159170.1 | - | 2 | 2886 | |||
BC081453.1 | - | 5 | 534 | 6820943 | ||
BC083135.1 | - | 5 | 497 | 6506450 | ||
BC085171.1 | - | 5 | 534 | 30699339 | ||
X56135.1 | - | 5 | 534 |