Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293652.1 | NP_001280581.1 | 8 | 1795 | ||
NM_001293653.1 | NP_001280582.1 | 6 | 1456 | |||
NM_001293654.1 | NP_001280583.1 | 7 | 1622 | |||
NM_001293655.1 | NP_001280584.1 | 6 | 1535 | |||
NM_009005.3 | NP_033031.2 | 6 | 1573 | |||
GenBank mRNA | AK005004.1 | - | 6 | 1536 | ||
AK057671.1 | - | 6 | 1660 | |||
AK135659.1 | - | 7 | 1625 | |||
AK148117.1 | - | 6 | 1567 | |||
AK154234.1 | - | 6 | 1554 | |||
AK158971.1 | - | 6 | 1457 | |||
AK159507.1 | - | 6 | 1502 | |||
AK159928.1 | - | 6 | 1474 | |||
BC004597.1 | - | 6 | 1491 | 3584891 | ||
BC086793.1 | - | 6 | 1489 | 6414012 | ||
X89650.1 | - | 6 | 1519 |