Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpjl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009036.1 | NP_033062.1 | 9 | 1085 | ||
GenBank mRNA | AK052449.1 | - | 7 | 4196 | ||
AK131814.1 | - | 9 | 1066 | |||
AK131832.1 | - | 9 | 1022 | |||
Y10926.1 | - | 9 | 1085 |