Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009042.1 | NP_033068.1 | 3 | 235 | ||
GenBank mRNA | AK075798.1 | - | 1 | 48 | ||
AK133506.1 | - | 3 | 239 | |||
AK160452.1 | - | 3 | 236 | |||
BC028761.1 | - | 3 | 231 | 1263580 |