Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009042.1 | NP_033068.1 | 6 | 647 | ||
GenBank mRNA | AK075798.1 | - | 4 | 460 | ||
AK133506.1 | - | 6 | 651 | |||
BC028761.1 | - | 6 | 643 | 1263580 |