Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009096.3 | NP_033122.1 | 3 | 241 | ||
GenBank mRNA | AK012284.1 | - | 2 | 211 | ||
AK012828.1 | - | 3 | 242 | |||
AK013333.1 | - | 2 | 212 | |||
AK013825.1 | - | 241 | ||||
AK028205.1 | - | 3 | 244 | |||
AK050609.1 | - | 3 | 242 | |||
AK146952.1 | - | 3 | 242 | |||
AK161991.1 | - | 3 | 244 | |||
AK166680.1 | - | 3 | 243 | |||
AK166727.1 | - | 3 | 287 | |||
AK168050.1 | - | 3 | 242 | |||
AK168496.1 | - | 3 | 221 | |||
AK169624.1 | - | 3 | 240 | |||
BC010604.1 | - | 3 | 227 | 3481640 | ||
BC090392.1 | - | 3 | 228 | 4459055 | ||
BC092050.1 | - | 3 | 228 | 3666815 | ||
Y00348.1 | - | 3 | 231 |