Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009125.2 | NP_033151.2 | 17, 18 | 2886 | ||
GenBank mRNA | AF041472.1 | - | 17, 18 | 2885 | ||
AK135879.1 | - | 7, 8 | 1047 | |||
AK135976.1 | - | 12, 13 | 1843 |