Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009128.2 | NP_033154.2 | 6 | 2165 | ||
GenBank mRNA | AK083085.1 | - | 6 | 1751 | ||
AK083922.1 | - | 6 | 1759 | |||
AK147406.1 | - | 6 | 2168 | |||
BC040384.1 | - | 6 | 1740 | 3155869 | ||
M26270.1 | - | 6 | 1753 |