Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009128.2 | NP_033154.2 | 6 | 4644 | ||
GenBank mRNA | AK078543.1 | - | 1 | 1050 | ||
AK147406.1 | - | 6 | 4647 | |||
AK159413.1 | - | 1 | 1362 | |||
BC040384.1 | - | 6 | 4219 | 3155869 | ||
M26270.1 | - | 6 | 4227 |