Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009129.2 | NP_033155.1 | 2 | 859 | ||
GenBank mRNA | BC014717.1 | - | 2 | 828 | ||
BC048249.1 | - | 2 | 759 | 6493827 |