Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164790.1 | NP_001158262.1 | 8, 9 | 1318 | ||
NM_009130.3 | NP_033156.1 | 8, 9 | 1318 | |||
GenBank mRNA | AK133655.1 | - | 8, 9 | 1220 | ||
AK139140.1 | - | 8, 9 | 1258 | |||
AK148498.1 | - | 8, 9 | 1302 | |||
AK158315.1 | - | 8, 9 | 1191 | |||
AK164089.1 | - | 8, 9 | 1261 | |||
BC024785.1 | - | 8, 9 | 1277 | 5362975 | ||
U02982.1 | - | 8, 9 | 1282 |