Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113478.1 | NP_001106950.1 | 3 | 324 | ||
NM_009146.3 | NP_033172.2 | 3 | 286 | |||
GenBank mRNA | AK075573.1 | - | 3 | 278 | ||
AK144933.1 | - | 3 | 278 | |||
AK162693.1 | - | 3 | 279 | |||
AK167289.1 | - | 3 | 314 | |||
AK210558.1 | - | 1 | 141 | |||
AU067508.1 | - | 3 | 286 | |||
BC027770.1 | - | 2 | 284 | 5344922 | ||
CN707391.1 | - | 3 | 324 | |||
D50464.1 | - | 3 | 274 |