Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113478.1 | NP_001106950.1 | 9 | 1154 | ||
NM_009146.3 | NP_033172.2 | 9 | 1116 | |||
GenBank mRNA | AK075573.1 | - | 9 | 1108 | ||
AK144933.1 | - | 9 | 1108 | |||
AK162693.1 | - | 9 | 1109 | |||
AK167289.1 | - | 9 | 1144 | |||
BC027770.1 | - | 8 | 1114 | 5344922 | ||
D50464.1 | - | 9 | 1104 |