Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGGTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209023.1 | - | 6 | 603 | ||
AK128861.1 | - | 5 | 3284 | |||
AK293057.1 | - | 9 | 876 | |||
BC020790.1 | - | 7 | 674 | 4290610 | ||
CR456941.1 | - | 6 | 603 | |||
U49245.1 | - | 6 | 603 | |||
X98001.1 | - | 6 | 627 | |||
Y08201.1 | - | 6 | 606 |