Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009177.4 | NP_033203.1 | 9 | 2019 | ||
GenBank mRNA | AK031344.1 | - | 9 | 1909 | ||
AK078469.1 | - | 8 | 1328 | |||
AK138747.1 | - | 9 | 1968 | |||
BC023130.1 | - | 8 | 2271 | |||
BC040763.1 | - | 6 | 1072 | |||
BC084730.1 | - | 6 | 1220 | 30355523 | ||
BC099670.1 | - | 9 | 1906 | 4187038 | ||
BC099693.1 | - | 10 | 1986 | 4910140 | ||
X73523.1 | - | 6 | 1037 |