Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289724.1 | NP_001276653.1 | 2, 3 | 491 | ||
NM_009193.2 | NP_033219.1 | 2, 3 | 491 | |||
GenBank mRNA | AK016826.1 | - | 1, 2 | 386 | ||
AK131729.1 | - | 262 | ||||
AK154053.1 | - | 2, 3 | 238 | |||
AK162121.1 | - | 2, 3 | 237 | |||
AK210098.1 | - | 1, 2 | 30 | |||
BC025113.1 | - | 2, 3 | 226 | |||
BC029631.1 | - | 2, 3 | 223 | 3500676 | ||
U75680.1 | - | 2, 3 | 223 |