Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289724.1 | NP_001276653.1 | 4 | 658 | ||
NM_001289725.1 | NP_001276654.1 | 4 | 613 | |||
NM_009193.2 | NP_033219.1 | 5 | 718 | |||
GenBank mRNA | AK131729.1 | - | 489 | |||
AK154053.1 | - | 5 | 465 | |||
AK162121.1 | - | 5 | 464 | |||
BC029631.1 | - | 4 | 390 | 3500676 | ||
U75680.1 | - | 5 | 450 |