Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009206.2 | NP_033232.2 | 4, 5 | 1078 | ||
GenBank mRNA | AF035526.1 | - | 5, 6 | 961 | ||
AK138398.1 | - | 4, 5 | 1033 | |||
AK147161.1 | - | 4, 5 | 416 | |||
AK160250.1 | - | 3, 4 | 953 | |||
BC072588.1 | - | 4, 5 | 1061 | |||
BC090964.1 | - | 4, 5 | 630 | 30625511 |