Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177792.1 | NP_001171263.1 | 8 | 696 | ||
NM_001177793.1 | NP_001171264.1 | 7 | 618 | |||
NM_009222.3 | NP_033248.1 | 7 | 663 | |||
GenBank mRNA | AB000822.1 | - | 6 | 538 | ||
AB007444.1 | - | 7 | 595 | |||
AF007169.1 | - | 6 | 487 | |||
AK017311.1 | - | 8 | 671 | |||
AK019162.1 | - | 7 | 664 | |||
BC070456.1 | - | 7 | 568 | 30606358 | ||
U73143.1 | - | 6 | 487 |