Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009272.4 | NP_033298.1 | 6, 7 | 1003 | ||
GenBank mRNA | AK028359.1 | - | 6, 7 | 1014 | ||
AK051101.1 | - | 6, 7 | 999 | |||
AK146395.1 | - | 6, 7 | 960 | |||
AK219795.1 | - | 14 | ||||
BC005566.1 | - | 6, 7 | 1007 | 3966710 | ||
L19311.1 | - | 6, 7 | 971 |