Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009274.2 | NP_033300.2 | 8, 9 | 980 | ||
GenBank mRNA | AB006036.1 | - | 8, 9 | 773 | ||
AK136861.1 | - | 8, 9 | 946 | |||
AK161384.1 | - | 9, 10 | 1039 | |||
AK169580.1 | - | 8, 9 | 926 | |||
AK196829.1 | - | 2, 3 | 96 | |||
BC020178.1 | - | 8, 9 | 877 | 4507346 | ||
U92456.1 | - | 9, 10 | 902 |
Gene Symbol: Asic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF410096.1 | - | 445 |