Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSSK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053006.4 | NP_443732.3 | 1 | 1625 | ||
GenBank mRNA | AF362953.1 | - | 1 | 1105 | ||
BC037781.2 | - | 1 | 1156 | 5273397 | ||
CR456587.1 | - | 1 | 1047 | |||
L77564.1 | - | 1 | 1625 |