Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205313.1 | NP_001192242.1 | 6, 7 | 757 | ||
NM_001205314.1 | NP_001192243.1 | 6, 7 | 793 | |||
NM_009283.4 | NP_033309.3 | 6, 7 | 808 | |||
GenBank mRNA | AK018544.1 | - | 6, 7 | 759 | ||
AK039458.1 | - | 6, 7 | 809 | |||
AK046517.1 | - | 6, 7 | 757 | |||
AK082706.1 | - | 6, 7 | 790 | |||
AK084855.1 | - | 6, 7 | 742 | |||
AK157263.1 | - | 6, 7 | 657 | |||
AK159887.1 | - | 6, 7 | 792 | |||
BC004808.1 | - | 6, 7 | 744 | 3587831 | ||
U06924.1 | - | 4, 5 | 462 |