Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164639.1 | NP_001158111.1 | 2 | 399 | ||
NM_009289.3 | NP_033315.2 | 2 | 399 | |||
GenBank mRNA | AF039574.1 | - | 2 | 404 | ||
AF112855.1 | - | 2 | 288 | |||
AK029491.1 | - | 2 | 266 | |||
AK122218.1 | - | 2 | 740 | |||
BC131674.1 | - | 2 | 300 | 40110915 | ||
BC131675.1 | - | 2 | 300 | 40110921 |