Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261419.1 | NP_001248348.1 | 4, 5 | 750 | ||
NM_004633.3 | NP_004624.1 | 4, 5 | 750 | |||
NR_048564.1 | 4, 5 | 737 | ||||
XM_006712734.3 | XP_006712797.1 | 4, 5 | 774 | |||
XM_006712736.3 | XP_006712799.1 | 4, 5 | 811 | |||
XM_011511801.2 | XP_011510103.1 | 4, 5 | 3110 | |||
XM_011511803.2 | XP_011510105.1 | 4, 5 | 2689 | |||
XM_011511804.2 | XP_011510106.1 | 4, 5 | 803 | |||
XM_011511805.2 | XP_011510107.1 | 4, 5 | 1576 | |||
XM_011511807.1 | XP_011510109.1 | 4, 5 | 3110 | |||
XM_017004889.1 | XP_016860378.1 | 3, 4 | 660 | |||
GenBank mRNA | AK130412.1 | - | 4, 5 | 634 | ||
BC027733.2 | - | 4, 5 | 704 | |||
BC039031.1 | - | 4, 5 | 750 | 5723005 | ||
FJ200480.1 | - | 3, 4 | 361 | |||
U64094.1 | - | 3, 4 | 582 | |||
X59770.1 | - | 3, 4 | 582 |