Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011077.2 | NP_035207.1 | 22 | 2842 | ||
GenBank mRNA | AK158481.1 | - | 22 | 2869 | ||
BC137925.1 | - | 22 | 2589 | 8860944 | ||
BC137926.1 | - | 22 | 2589 | 8860945 | ||
EF194891.1 | - | 22 | 2376 | |||
U49908.1 | - | 22 | 2834 | |||
U73910.1 | - | 22 | 2842 | |||
U75646.1 | - | 22 | 2423 |